siRNA Details
| Gene Symbol: C14orf79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174891.3 | NP_777551.2 | 5 | 1084 | ||
| GenBank mRNA | AF038188.1 | - | 5 | 648 | ||
| AK057249.1 | - | 6 | 1160 | |||
| BC003539.1 | - | 4 | 396 | |||
| BC011537.1 | - | 4 | 914 | |||
| BC025175.1 | - | 5 | 595 | |||
| BC137495.1 | - | 5 | 1027 | 9021495 | ||
| BC144671.1 | - | 5 | 1013 | |||

C14orf79
Genomic Map