siRNA Details
| Gene Symbol: C14orf79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174891.3 | NP_777551.2 | 5 | 1852 | ||
| GenBank mRNA | AF038188.1 | - | 5 | 1416 | ||
| AK057249.1 | - | 6 | 1928 | |||
| BC003539.1 | - | 4 | 1164 | |||
| BC011537.1 | - | 4 | 1682 | |||
| BC025175.1 | - | 5 | 1363 | |||

C14orf79
Genomic Map