siRNA Details
Gene Symbol: NLRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317000.1 | NP_001303929.1 | 1 | 328 | ||
NM_176811.2 | NP_789781.2 | 1 | 328 | |||
GenBank mRNA | AY154463.1 | - | 1 | 257 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NLRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317000.1 | NP_001303929.1 | 1 | 328 | ||
NM_176811.2 | NP_789781.2 | 1 | 328 | |||
GenBank mRNA | AY154463.1 | - | 1 | 257 |