siRNA Details
| Gene Symbol: CHMP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176812.4 | NP_789782.1 | 3 | 615 | ||
| GenBank mRNA | AB100261.1 | - | 3 | 560 | ||
| AY329085.1 | - | 3 | 450 | |||
| AY726594.1 | - | 3 | 462 | |||
| BC033859.1 | - | 3 | 559 | 5173394 | ||
| BQ926070.1 | - | 609 | ||||

CHMP4B
Genomic Map