siRNA Details
| Gene Symbol: TAS2R40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176882.1 | NP_795363.1 | 1 | 415 | ||
| GenBank mRNA | BC101795.1 | - | 1 | 457 | 8069301 | |
| BC101797.1 | - | 1 | 457 | 8069303 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS2R40| Gene Symbol: TAS2R40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176882.1 | NP_795363.1 | 1 | 415 | ||
| GenBank mRNA | BC101795.1 | - | 1 | 457 | 8069301 | |
| BC101797.1 | - | 1 | 457 | 8069303 | ||