siRNA Details
| Gene Symbol: TRAPPC6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079537.1 | NP_001073005.1 | 6 | 2040 | ||
| NM_177452.3 | NP_803235.1 | 5 | 1956 | |||
| XM_011536409.2 | XP_011534711.1 | 7 | 2060 | |||
| GenBank mRNA | AK025437.1 | - | 1 | 698 | ||
| AK056690.1 | - | 5 | 1926 | |||
| AL833179.1 | - | 6 | 1924 | |||
| BC047328.1 | - | 5 | 1840 | 5534151 | ||
| BX161510.1 | - | 6 | 1932 | |||

TRAPPC6B
Genomic Map