siRNA Details
Gene Symbol: OTOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177998.1 | NP_819056.1 | 2 | 537 | ||
GenBank mRNA | BC130430.1 | - | 2 | 532 | 40146461 | |
BC130432.1 | - | 2 | 532 | 40146463 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OTOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177998.1 | NP_819056.1 | 2 | 537 | ||
GenBank mRNA | BC130430.1 | - | 2 | 532 | 40146461 | |
BC130432.1 | - | 2 | 532 | 40146463 |