siRNA Details
| Gene Symbol: LRRTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178011.4 | NP_821079.3 | 3 | 3562 | ||
| NR_111909.1 | 3 | 1606 | ||||
| GenBank mRNA | BC111492.1 | - | 3 | 3514 | 5196989 | |
| BX640611.1 | - | 3 | 3540 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRTM3| Gene Symbol: LRRTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178011.4 | NP_821079.3 | 3 | 3562 | ||
| NR_111909.1 | 3 | 1606 | ||||
| GenBank mRNA | BC111492.1 | - | 3 | 3514 | 5196989 | |
| BX640611.1 | - | 3 | 3540 | |||