siRNA Details
| Gene Symbol: NEK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178170.2 | NP_835464.1 | 11 | 1502 | ||
| XM_011524638.2 | XP_011522940.1 | 12 | 1745 | |||
| XM_011524640.2 | XP_011522942.1 | 12 | 1706 | |||
| XM_017024499.1 | XP_016879988.1 | 11 | 1551 | |||
| XM_017024500.1 | XP_016879989.1 | 11 | 1587 | |||
| GenBank mRNA | AL833909.1 | - | 2 | 172 | ||
| AY242354.1 | - | 11 | 1502 | |||
| AY267371.1 | - | 11 | 1502 | |||
| BC112240.1 | - | 11 | 1536 | 8327708 | ||
| BC113705.1 | - | 11 | 1536 | 8322757 | ||

NEK8
Genomic Map