siRNA Details
| Gene Symbol: GSDMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178171.4 | NP_835465.2 | 10, 11 | 1138 | ||
| XM_006721832.2 | XP_006721895.1 | 10, 11 | 1091 | |||
| XM_011524651.2 | XP_011522953.1 | 8, 9 | 854 | |||
| XM_017024502.1 | XP_016879991.1 | 9, 10 | 1111 | |||
| GenBank mRNA | AB093591.1 | - | 9, 10 | 1031 | ||
| AF307953.1 | - | 9, 10 | 1053 | |||
| AK096439.1 | - | 11, 12 | 1140 | |||
| BC109197.2 | - | 11, 12 | 1100 | 40005562 | ||

GSDMA
Genomic Map