siRNA Details
| Gene Symbol: GSDMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178171.4 | NP_835465.2 | 12 | 1232 | ||
| XM_006721832.2 | XP_006721895.1 | 12 | 1185 | |||
| XM_011524651.2 | XP_011522953.1 | 10 | 948 | |||
| XM_017024502.1 | XP_016879991.1 | 11 | 1205 | |||
| GenBank mRNA | AB093591.1 | - | 11 | 1125 | ||
| AF307953.1 | - | 11 | 1147 | |||
| AK096439.1 | - | 13 | 1234 | |||
| BC109197.2 | - | 13 | 1194 | 40005562 | ||

GSDMA
Genomic Map