siRNA Details
| Gene Symbol: MSS51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024593.1 | NP_001019764.1 | 7 | 2346 | ||
| GenBank mRNA | AK096884.1 | - | 7 | 1917 | ||
| BC094693.1 | - | 7 | 2325 | 30529259 | ||
| BX537678.1 | - | 5 | 3471 | |||
| CR936824.1 | - | 6 | 2653 | |||

MSS51
Genomic Map