siRNA Details
| Gene Symbol: ZDHHC21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178566.4 | NP_848661.1 | 8 | 1036 | ||
| XM_017014682.1 | XP_016870171.1 | 6 | 820 | |||
| XM_017014683.1 | XP_016870172.1 | 9 | 1076 | |||
| XM_017014684.1 | XP_016870173.1 | 9 | 1008 | |||
| XM_017014685.1 | XP_016870174.1 | 9 | 956 | |||
| XM_017014686.1 | XP_016870175.1 | 10 | 1137 | |||
| XM_017014687.1 | XP_016870176.1 | 9 | 1084 | |||
| XM_017014688.1 | XP_016870177.1 | 9 | 1166 | |||
| XM_017014689.1 | XP_016870178.1 | 9 | 1142 | |||
| XM_017014690.1 | XP_016870179.1 | 10 | 1026 | |||
| XM_017014691.1 | XP_016870180.1 | 9 | 3551 | |||
| XM_017014692.1 | XP_016870181.1 | 9 | 816 | |||
| XM_017014693.1 | XP_016870182.1 | 10 | 873 | |||
| GenBank mRNA | AK096860.1 | - | 1 | 41 | ||
| AK127313.1 | - | 1 | 1299 | |||
| AK292960.1 | - | 8 | 843 | |||
| BC042558.1 | - | 8 | 1030 | 4837003 | ||

ZDHHC21
Genomic Map