siRNA Details
Gene Symbol: RNF113B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC017585.2 | - | 2 | 1273 | 4828542 | |
BC025388.1 | - | 2 | 1266 | 4824788 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RNF113B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC017585.2 | - | 2 | 1273 | 4828542 | |
BC025388.1 | - | 2 | 1266 | 4824788 |