siRNA Details
| Gene Symbol: MRPL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181514.1 | NP_852615.1 | 4 | 279 | ||
| NM_181515.1 | NP_852616.1 | 4 | 292 | |||
| XM_005273823.4 | XP_005273880.1 | 4 | 276 | |||
| GenBank mRNA | AK096756.1 | - | 4 | 292 | ||
| AK301989.1 | - | 4 | 276 | |||
| AV711404.1 | - | 4 | 151 | |||
| BC055088.1 | - | 4 | 278 | 6457142 | ||
| BM916618.1 | - | 4 | 244 | |||

MRPL21
Genomic Map