siRNA Details
| Gene Symbol: THAP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130475.2 | NP_001123947.1 | 3 | 762 | ||
| NM_001287598.1 | NP_001274527.1 | 3 | 716 | |||
| NM_001287599.1 | NP_001274528.1 | 3 | 640 | |||
| NM_001287601.1 | NP_001274530.1 | 2 | 569 | |||
| NM_182529.3 | NP_872335.2 | 3 | 838 | |||
| GenBank mRNA | AL833137.1 | - | 3 | 717 | ||
| BC053634.1 | - | 3 | 838 | 5165075 | ||
| BF244164.1 | - | 3 | 483 | |||
| BI830307.1 | - | 3 | 623 | |||

THAP5
Genomic Map