siRNA Details
| Gene Symbol: RNF144B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182757.3 | NP_877434.2 | 8 | 1685 | ||
| XM_005248986.3 | XP_005249043.1 | 7 | 1601 | |||
| GenBank mRNA | AB076367.1 | - | 8 | 1669 | ||
| AK096832.1 | - | 8 | 1685 | |||
| AL832329.1 | - | 8 | 1510 | |||
| BC063311.1 | - | 8 | 1556 | 30331343 | ||

RNF144B
Genomic Map