siRNA Details
Gene Symbol: GDF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182828.3 | NP_878248.2 | 2 | 1411 | ||
GenBank mRNA | AB158468.1 | - | 2 | 893 | ||
AF522369.1 | - | 2 | 1411 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GDF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182828.3 | NP_878248.2 | 2 | 1411 | ||
GenBank mRNA | AB158468.1 | - | 2 | 893 | ||
AF522369.1 | - | 2 | 1411 |