siRNA Details
| Gene Symbol: JAKMIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105521.3 | NP_001098991.1 | 24 | 4233 | ||
| XM_017016095.1 | XP_016871584.1 | 24 | 6148 | |||
| GenBank mRNA | AK126964.1 | - | 14 | 2520 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

JAKMIP3| Gene Symbol: JAKMIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105521.3 | NP_001098991.1 | 24 | 4233 | ||
| XM_017016095.1 | XP_016871584.1 | 24 | 6148 | |||
| GenBank mRNA | AK126964.1 | - | 14 | 2520 | ||