siRNA Details
| Gene Symbol: PRICKLE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198859.3 | NP_942559.1 | 8 | 3474 | ||
| XM_011533432.2 | XP_011531734.1 | 10 | 3358 | |||
| XM_011533433.2 | XP_011531735.1 | 9 | 3268 | |||
| XM_011533434.2 | XP_011531736.1 | 10 | 3183 | |||
| XM_011533435.2 | XP_011531737.1 | 9 | 3093 | |||
| XM_011533436.2 | XP_011531738.1 | 9 | 3416 | |||
| XM_011533437.2 | XP_011531739.1 | 9 | 12486 | |||
| XM_011533438.2 | XP_011531740.1 | 6 | 3308 | |||
| XM_011533440.2 | XP_011531742.1 | 10 | 3288 | |||
| XM_017005798.1 | XP_016861287.1 | 8 | 3198 | |||
| XM_017005799.1 | XP_016861288.1 | 7 | 4947 | |||
| GenBank mRNA | AK127113.1 | - | 1 | 1489 | ||
| AK127839.1 | - | 1 | 605 | |||
| AL833539.1 | - | 8 | 3475 | |||
| BC040867.1 | - | 2 | 1330 | |||
| BC119002.1 | - | 8 | 3455 | 40082599 | ||
| BX537915.1 | - | 2 | 1302 | |||
| BX647797.1 | - | 3 | 2171 | |||
| BX649125.1 | - | 1 | 614 | |||
| CR936678.1 | - | 2 | 1524 | |||

PRICKLE2
Genomic Map