siRNA Details
| Gene Symbol: WNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282394.1 | NP_001269323.1 | 26 | 6271 | ||
| NM_006648.3 | NP_006639.3 | 25 | 6160 | |||
| XM_005252137.3 | XP_005252194.1 | 26 | 6392 | |||
| XM_005252140.2 | XP_005252197.1 | 26 | 6392 | |||
| XM_005252141.2 | XP_005252198.1 | 26 | 6392 | |||
| XM_005252144.2 | XP_005252201.1 | 26 | 6392 | |||
| XM_011518926.2 | XP_011517228.1 | 26 | 6392 | |||
| XM_011518927.2 | XP_011517229.1 | 26 | 6389 | |||
| XM_011518928.2 | XP_011517230.1 | 26 | 6338 | |||
| XM_011518930.2 | XP_011517232.1 | 26 | 6257 | |||
| XM_011518932.2 | XP_011517234.1 | 26 | 6392 | |||
| XM_011518937.2 | XP_011517239.1 | 25 | 5700 | |||
| XM_011518939.2 | XP_011517241.1 | 25 | 5745 | |||
| XM_011518940.2 | XP_011517242.1 | 25 | 5747 | |||
| XM_017015045.1 | XP_016870534.1 | 26 | 6389 | |||
| XM_017015046.1 | XP_016870535.1 | 26 | 6254 | |||
| XM_017015047.1 | XP_016870536.1 | 26 | 6338 | |||
| XM_017015048.1 | XP_016870537.1 | 26 | 6335 | |||
| XM_017015051.1 | XP_016870540.1 | 26 | 6257 | |||
| GenBank mRNA | AB051547.2 | - | 25 | 6147 | ||
| AJ242724.4 | - | 26 | 6271 | |||
| AK000694.1 | - | 11 | 2550 | |||
| BC037965.1 | - | 7 | 1459 | |||

WNK2
Genomic Map