siRNA Details
| Gene Symbol: NAV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024383.1 | NP_001019554.1 | 6 | 860 | ||
| NM_014903.5 | NP_055718.4 | 6 | 860 | |||
| XM_005269215.3 | XP_005269272.1 | 6 | 4861 | |||
| XM_011538940.2 | XP_011537242.1 | 6 | 4862 | |||
| XM_011538941.2 | XP_011537243.1 | 6 | 4861 | |||
| XM_011538944.2 | XP_011537246.1 | 6 | 4859 | |||
| XM_017020164.1 | XP_016875653.1 | 6 | 4863 | |||
| XM_017020165.1 | XP_016875654.1 | 6 | 4863 | |||
| XM_017020166.1 | XP_016875655.1 | 6 | 849 | |||
| XM_017020168.1 | XP_016875657.1 | 6 | 4862 | |||
| XM_017020169.1 | XP_016875658.1 | 6 | 4861 | |||
| XM_017020170.1 | XP_016875659.1 | 6 | 4858 | |||
| GenBank mRNA | AF397731.1 | - | 5 | 444 | ||
| AJ488201.1 | - | 6 | 732 | |||
| BC017667.2 | - | 6 | 1237 | |||

NAV3
Genomic Map