siRNA Details
| Gene Symbol: WNT8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003393.3 | NP_003384.2 | 2, 3 | 224 | ||
| GenBank mRNA | AB073637.1 | - | 2, 3 | 222 | ||
| CN276452.1 | - | 2, 3 | 224 | |||
| X91940.1 | - | 1, 2 | 44 | |||
| Y11094.1 | - | 2, 3 | 247 | |||

WNT8B
Genomic Map