siRNA Details
| Gene Symbol: LRRC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022143.4 | NP_071426.1 | 2 | 2347 | ||
| XM_011516461.2 | XP_011514763.1 | 3 | 2653 | |||
| XM_017012505.1 | XP_016867994.1 | 3 | 2543 | |||
| GenBank mRNA | AF196976.4 | - | 2 | 2350 | ||
| AK172751.1 | - | 2 | 2351 | |||
| BC036685.1 | - | 1 | 21 | |||
| BC111561.1 | - | 2 | 2341 | 40068142 | ||
| BC111745.1 | - | 2 | 2341 | 40068146 | ||

LRRC4
Genomic Map