siRNA Details
| Gene Symbol: MOAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022151.4 | NP_071434.2 | 3 | 1131 | ||
| GenBank mRNA | AF305550.1 | - | 1 | 761 | ||
| AK024029.1 | - | 3 | 1103 | |||
| AK315522.1 | - | 3 | 1143 | |||
| BC015044.1 | - | 3 | 1065 | 3922055 | ||

MOAP1
Genomic Map