siRNA Details
| Gene Symbol: RBSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302378.1 | NP_001289307.1 | 5 | 806 | ||
| NM_022340.3 | NP_071735.2 | 6 | 962 | |||
| XM_005265384.4 | XP_005265441.1 | 6 | 925 | |||
| XM_005265385.4 | XP_005265442.1 | 5 | 780 | |||
| XM_011534001.2 | XP_011532303.1 | 2 | 302 | |||
| XM_017007023.1 | XP_016862512.1 | 5 | 798 | |||
| XM_017007024.1 | XP_016862513.1 | 7 | 1108 | |||
| XM_017007025.1 | XP_016862514.1 | 6 | 967 | |||
| XM_017007026.1 | XP_016862515.1 | 5 | 812 | |||
| XM_017007027.1 | XP_016862516.1 | 6 | 953 | |||
| XM_017007028.1 | XP_016862517.1 | 6 | 953 | |||
| GenBank mRNA | AB209673.1 | - | 6 | 922 | ||
| AK291288.1 | - | 5 | 797 | |||
| AK301735.1 | - | 5 | 798 | |||
| AL833545.1 | - | 6 | 956 | |||
| AY009133.1 | - | 3 | 339 | |||
| BC106940.2 | - | 3 | 339 | 40034008 | ||

RBSN
Genomic Map