siRNA Details
| Gene Symbol: NOC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022451.10 | NP_071896.8 | 2 | 327 | ||
| GenBank mRNA | AB077992.1 | - | 2 | 214 | ||
| AK091246.1 | - | 2 | 284 | |||
| AK292520.1 | - | 2 | 309 | |||
| BC049850.1 | - | 2 | 277 | 5549517 | ||
| BX505710.1 | - | 2 | 329 | |||

NOC3L
Genomic Map