siRNA Details
| Gene Symbol: CDH24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022478.3 | NP_071923.2 | 5, 6 | 1053 | ||
| NM_144985.3 | NP_659422.2 | 5, 6 | 1053 | |||
| XM_011537089.1 | XP_011535391.1 | 5, 6 | 1134 | |||
| GenBank mRNA | AK057922.1 | - | 5, 6 | 989 | ||
| AK289576.1 | - | 5, 6 | 1053 | |||
| AK307746.1 | - | 5, 6 | 1034 | |||
| AY260900.1 | - | 4, 5 | 886 | |||
| AY260901.1 | - | 4, 5 | 886 | |||
| AY358199.1 | - | 5, 6 | 1009 | |||
| BX248303.1 | - | 1, 2 | 77 | |||

CDH24
Genomic Map