siRNA Details
| Gene Symbol: PCNX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022495.5 | NP_071940.4 | 9 | 2997 | ||
| XM_005268009.2 | XP_005268066.1 | 10 | 4374 | |||
| GenBank mRNA | AK022861.1 | - | 2 | 455 | ||
| AK023008.1 | - | 4 | 1485 | |||
| AK095489.1 | - | 9 | 2981 | |||
| AL136581.1 | - | 2 | 515 | |||
| AY553876.1 | - | 2 | 498 | |||
| BC033843.1 | - | 2 | 982 | |||
| BX648440.1 | - | 5 | 1721 | |||

PCNX4
Genomic Map