siRNA Details
| Gene Symbol: ALDH8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193480.1 | NP_001180409.1 | 6 | 1483 | ||
| NM_022568.3 | NP_072090.1 | 7 | 1633 | |||
| NM_170771.2 | NP_739577.1 | 6 | 1471 | |||
| GenBank mRNA | AK074266.1 | - | 6 | 1464 | ||
| AK298325.1 | - | 6 | 1445 | |||
| BC113862.1 | - | 7 | 1603 | 40033493 | ||
| BC114473.1 | - | 7 | 1479 | 40033488 | ||
| BX647324.1 | - | 1 | 1214 | |||

ALDH8A1
Genomic Map