siRNA Details
| Gene Symbol: PARP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022750.3 | NP_073587.1 | 7, 8 | 2206 | ||
| XM_005250038.3 | XP_005250095.1 | 7, 8 | 2201 | |||
| XM_011516491.2 | XP_011514793.1 | 7, 8 | 2201 | |||
| GenBank mRNA | AL136766.1 | - | 7, 8 | 1828 | ||
| AL137255.1 | - | 7, 8 | 1025 | |||
| BC081541.1 | - | 7, 8 | 2206 | 5296560 | ||

PARP12
Genomic Map