siRNA Details
| Gene Symbol: ZNF559-ZNF177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172650.2 | NP_001166121.1 | 10 | 947 | ||
| NM_001202425.1 | NP_001189354.1 | 7 | 680 | |||
| GenBank mRNA | U37251.1 | - | 7 | 650 | ||
| Gene Symbol: ZNF177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172651.1 | NP_001166122.1 | 4 | 364 | ||
| NM_003451.2 | NP_003442.2 | 7 | 659 | |||
| GenBank mRNA | AK302274.1 | - | 3 | 269 | ||
| BC012012.1 | - | 3 | 250 | 4509206 | ||
| BP260204.1 | - | 2 | 150 | |||
| U37263.1 | - | 7 | 659 | |||

ZNF559-ZNF177
Genomic Map