siRNA Details
| Gene Symbol: ZNF205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042428.1 | NP_001035893.1 | 3 | 386 | ||
| NM_001278158.1 | NP_001265087.1 | 3 | 705 | |||
| NM_003456.2 | NP_003447.2 | 3 | 454 | |||
| XM_005255558.2 | XP_005255615.1 | 3 | 421 | |||
| GenBank mRNA | AB672633.1 | - | 3 | 698 | ||
| AF060865.1 | - | 3 | 411 | |||
| AK313009.1 | - | 3 | 443 | |||
| BC002810.2 | - | 3 | 411 | 3635500 | ||
| CB152017.1 | - | 3 | 454 | |||

ZNF205
Genomic Map