siRNA Details
| Gene Symbol: GGCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199815.1 | NP_001186744.1 | 4 | 1175 | ||
| NM_001199816.1 | NP_001186745.1 | 3 | 1019 | |||
| NM_001199817.1 | NP_001186746.1 | 2 | 873 | |||
| NM_024051.3 | NP_076956.1 | 4 | 1155 | |||
| NR_037669.1 | 3 | 1009 | ||||
| GenBank mRNA | AK021779.1 | - | 3 | 1634 | ||
| BC000625.2 | - | 4 | 1150 | 3346822 | ||
| BC005356.1 | - | 3 | 1007 | 4102029 | ||
| BC013937.1 | - | 4 | 1116 | 3997629 | ||
| BC019243.1 | - | 4 | 1147 | 3828006 | ||

GGCT
Genomic Map