siRNA Details
| Gene Symbol: GID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024052.4 | NP_076957.3 | 5 | 986 | ||
| GenBank mRNA | AK127580.1 | - | 5 | 673 | ||
| AK315083.1 | - | 5 | 874 | |||
| BC000636.2 | - | 5 | 647 | |||
| BC041829.1 | - | 5 | 991 | 5268071 | ||

GID4
Genomic Map