siRNA Details
| Gene Symbol: GID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024052.4 | NP_076957.3 | 6 | 1324 | ||
| XM_005256800.1 | XP_005256857.1 | 5 | 1080 | |||
| GenBank mRNA | AK074284.1 | - | 6 | 1199 | ||
| AK127580.1 | - | 6 | 1011 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GID4| Gene Symbol: GID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024052.4 | NP_076957.3 | 6 | 1324 | ||
| XM_005256800.1 | XP_005256857.1 | 5 | 1080 | |||
| GenBank mRNA | AK074284.1 | - | 6 | 1199 | ||
| AK127580.1 | - | 6 | 1011 | |||