siRNA Details
| Gene Symbol: KCTD15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024076.2 | NP_076981.2 | 7 | 1251 | ||
| GenBank mRNA | BC001185.1 | - | 7 | 1235 | 3502748 | |
| BC009335.2 | - | 7 | 1235 | 4128019 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCTD15| Gene Symbol: KCTD15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024076.2 | NP_076981.2 | 7 | 1251 | ||
| GenBank mRNA | BC001185.1 | - | 7 | 1235 | 3502748 | |
| BC009335.2 | - | 7 | 1235 | 4128019 | ||