siRNA Details
| Gene Symbol: KDELC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318732.1 | NP_001305661.1 | 2 | 629 | ||
| NM_024089.2 | NP_076994.2 | 2 | 629 | |||
| XM_005254075.2 | XP_005254132.1 | 2 | 629 | |||
| GenBank mRNA | AK074974.1 | - | 2 | 547 | ||
| AK222567.1 | - | 2 | 551 | |||
| AK225103.1 | - | 2 | 552 | |||
| AK309238.1 | - | 2 | 552 | |||
| AY358959.1 | - | 2 | 530 | |||
| BC001297.1 | - | 2 | 582 | 3456869 | ||

KDELC1
Genomic Map