siRNA Details
| Gene Symbol: FKRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039885.2 | NP_001034974.1 | 4 | 2034 | ||
| NM_024301.4 | NP_077277.1 | 4 | 1982 | |||
| XM_005259247.2 | XP_005259304.1 | 5 | 2095 | |||
| XM_005259248.2 | XP_005259305.1 | 5 | 2085 | |||
| XM_005259249.4 | XP_005259306.1 | 5 | 2027 | |||
| XM_011527306.1 | XP_011525608.1 | 3 | 1831 | |||
| XM_011527307.1 | XP_011525609.1 | 4 | 1880 | |||
| XM_017027297.1 | XP_016882786.1 | 4 | 2201 | |||
| GenBank mRNA | AJ314847.1 | - | 4 | 1970 | ||
| AK022638.1 | - | 4 | 1970 | |||
| AK095497.1 | - | 4 | 2021 | |||
| AK291282.1 | - | 4 | 1970 | |||
| BC002612.2 | - | 2 | 1778 | 3160297 | ||

FKRP
Genomic Map