siRNA Details
| Gene Symbol: LRRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024512.4 | NP_078788.2 | 9 | 1629 | ||
| XM_011534110.2 | XP_011532412.1 | 9 | 1418 | |||
| GenBank mRNA | AJ308569.2 | - | 9 | 1403 | ||
| AK057808.1 | - | 9 | 1471 | |||
| AK223440.1 | - | 9 | 1471 | |||
| BC029118.1 | - | 9 | 1401 | 5183639 | ||

LRRC2
Genomic Map