siRNA Details
| Gene Symbol: GCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024523.5 | NP_078799.2 | 2 | 3319 | ||
| GenBank mRNA | AK025688.1 | - | 2 | 1978 | ||
| AK092878.1 | - | 2 | 2775 | |||
| AL832833.1 | - | 2 | 1981 | |||
| BC008902.2 | - | 2 | 2101 | |||
| BC014100.2 | - | 2 | 3319 | 2988591 | ||
| BC078665.1 | - | 2 | 3271 | 5923117 | ||

GCC1
Genomic Map