siRNA Details
Gene Symbol: TTC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122835.2 | NP_001116307.2 | 17 | 2029 | ||
NM_024525.4 | NP_078801.3 | 19 | 2191 | |||
XM_005273260.1 | XP_005273317.1 | 19 | 2188 | |||
XM_005273261.2 | XP_005273318.1 | 18 | 2098 | |||
XM_005273262.1 | XP_005273319.1 | 17 | 2032 | |||
XM_005273264.2 | XP_005273321.1 | 17 | 1963 | |||
XM_006711814.2 | XP_006711877.1 | 15 | 1804 | |||
XM_006711815.2 | XP_006711878.1 | 12 | 3198 | |||
XM_011544275.2 | XP_011542577.1 | 16 | 1870 | |||
XM_011544276.2 | XP_011542578.1 | 12 | 1434 | |||
XM_017002321.1 | XP_016857810.1 | 18 | 2095 | |||
XM_017002322.1 | XP_016857811.1 | 17 | 1960 | |||
XM_017002323.1 | XP_016857812.1 | 15 | 1766 | |||
XM_017002324.1 | XP_016857813.1 | 15 | 1801 | |||
XM_017002325.1 | XP_016857814.1 | 19 | 5393 | |||
XM_017002326.1 | XP_016857815.1 | 14 | 4773 | |||
GenBank mRNA | AK026237.1 | - | 6 | 498 | ||
AK075386.1 | - | 19 | 2157 | |||
AK075561.1 | - | 17 | 2008 | |||
AK309725.1 | - | 15 | 1847 | |||
AL834453.1 | - | 10 | 1157 | |||
BC042683.1 | - | 10 | 1075 |
