siRNA Details
| Gene Symbol: VCPKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040662.1 | NP_001035752.1 | 2 | 319 | ||
| NM_024558.2 | NP_078834.2 | 2 | 319 | |||
| NR_049738.1 | 2 | 319 | ||||
| NR_049739.1 | 1 | 426 | ||||
| XM_005268060.3 | XP_005268117.1 | 2 | 305 | |||
| XM_017021640.1 | XP_016877129.1 | 2 | 305 | |||
| XM_017021641.1 | XP_016877130.1 | 2 | 305 | |||
| XM_017021642.1 | XP_016877131.1 | 1 | 402 | |||
| GenBank mRNA | AK023982.1 | - | 2 | 319 | ||
| AK129564.1 | - | 1 | 305 | |||
| BC105118.1 | - | 2 | 340 | 8144121 | ||
| BC143673.1 | - | 2 | 340 | 9052187 | ||
| BC143674.1 | - | 2 | 340 | 9052188 | ||
| BI753530.1 | - | 2 | 299 | |||
| BI767189.1 | - | 2 | 290 | |||
| BX247997.1 | - | 1 | 405 | |||

VCPKMT
Genomic Map