siRNA Details
| Gene Symbol: ARMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286562.1 | NP_001273491.1 | 3 | 625 | ||
| NM_024573.2 | NP_078849.1 | 4 | 731 | |||
| GenBank mRNA | AK022972.1 | - | 4 | 702 | ||
| AK298490.1 | - | 3 | 412 | |||
| AK310840.1 | - | 3 | 389 | |||
| BC011348.1 | - | 4 | 451 | 4340090 | ||

ARMT1
Genomic Map