siRNA Details
| Gene Symbol: FAM184A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100411.2 | NP_001093881.1 | 2 | 403 | ||
| NM_001288576.1 | NP_001275505.1 | 2 | 403 | |||
| NM_024581.5 | NP_078857.5 | 2 | 794 | |||
| GenBank mRNA | AK091682.1 | - | 2 | 399 | ||
| AK308316.1 | - | 2 | 890 | |||
| BC060769.1 | - | 2 | 794 | 30337859 | ||
| BX538186.1 | - | 2 | 383 | |||
| BX640728.1 | - | 2 | 452 | |||
| DA717412.1 | - | 2 | 403 | |||

FAM184A
Genomic Map