siRNA Details
| Gene Symbol: PLEKHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024613.3 | NP_078889.1 | 2 | 1375 | ||
| GenBank mRNA | AK023249.1 | - | 2 | 1148 | ||
| AL834473.1 | - | 2 | 1298 | |||
| BC011806.2 | - | 2 | 1275 | 4335031 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLEKHF2| Gene Symbol: PLEKHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024613.3 | NP_078889.1 | 2 | 1375 | ||
| GenBank mRNA | AK023249.1 | - | 2 | 1148 | ||
| AL834473.1 | - | 2 | 1298 | |||
| BC011806.2 | - | 2 | 1275 | 4335031 | ||