siRNA Details
| Gene Symbol: MANEA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024641.3 | NP_078917.2 | 5 | 1444 | ||
| XM_005267147.3 | XP_005267204.1 | 5 | 1837 | |||
| GenBank mRNA | AJ577574.1 | - | 4 | 1309 | ||
| AK022900.1 | - | 4 | 950 | |||
| AK291940.1 | - | 5 | 1375 | |||
| AY372528.1 | - | 4 | 1264 | |||
| BC137014.1 | - | 4 | 1313 | 9021007 | ||
| BC137016.1 | - | 4 | 1313 | 9021009 | ||
| BC146671.1 | - | 5 | 1379 | 40148201 | ||
| BC150199.1 | - | 5 | 1379 | 40148811 | ||
| BX537398.1 | - | 5 | 1375 | |||
| BX640869.1 | - | 5 | 1405 | |||
| CR936827.1 | - | 5 | 1379 | |||

MANEA
Genomic Map