siRNA Details
| Gene Symbol: ZC2HC1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026746.1 | - | 2 | 223 | ||
| AK303882.1 | - | 2 | 226 | |||
| HY031397.1 | - | 2 | 288 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZC2HC1C| Gene Symbol: ZC2HC1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026746.1 | - | 2 | 223 | ||
| AK303882.1 | - | 2 | 226 | |||
| HY031397.1 | - | 2 | 288 | |||