siRNA Details
| Gene Symbol: THAP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317776.1 | NP_001304705.1 | 4 | 593 | ||
| NM_024672.5 | NP_078948.3 | 3 | 527 | |||
| XM_005263220.2 | XP_005263277.1 | 3 | 484 | |||
| XM_011532246.2 | XP_011530548.1 | 2 | 202 | |||
| XM_017008603.1 | XP_016864092.1 | 3 | 1367 | |||
| XM_017008604.1 | XP_016864093.1 | 3 | 1420 | |||
| XM_017008605.1 | XP_016864094.1 | 3 | 461 | |||
| XM_017008606.1 | XP_016864095.1 | 4 | 585 | |||
| XM_017008607.1 | XP_016864096.1 | 3 | 800 | |||
| XM_017008608.1 | XP_016864097.1 | 4 | 1439 | |||
| XM_017008609.1 | XP_016864098.1 | 3 | 474 | |||
| XM_017008610.1 | XP_016864099.1 | 3 | 461 | |||
| GenBank mRNA | AK091412.1 | - | 3 | 424 | ||
| BC131722.1 | - | 4 | 474 | |||
| BX648134.1 | - | 3 | 632 | |||
| DA317935.1 | - | 3 | 493 | |||
| DB093127.1 | - | 3 | 528 | |||

THAP9
Genomic Map