siRNA Details
| Gene Symbol: TST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270483.1 | NP_001257412.1 | 3 | 1028 | ||
| NM_003312.5 | NP_003303.2 | 3 | 1060 | |||
| GenBank mRNA | AK000862.1 | - | 3 | 1054 | ||
| AK091874.1 | - | 2 | 1680 | |||
| AK223521.1 | - | 3 | 1054 | |||
| AK301300.1 | - | 4 | 895 | |||
| BC010148.2 | - | 3 | 1037 | 4329727 | ||
| CR456598.1 | - | 2 | 1063 | |||

TST
Genomic Map